ENA018108500 Enamine

3-(2,3-dihydro-1,4-benzodioxine-6-sulfonamido)propanoic acid

  • MDL number MFCD00706444
  • Cas Number : 306278-42-2
  • Molecular Weight : 287.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-166
Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05195

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05195
Availability details for
Quantity
  • Availability
  • Add to Cart