ENA018108512 Enamine

4-(2,5-difluorophenyl)-1,3-thiazol-2-amine

  • MDL number MFCD02663893
  • Cas Number : 383132-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 147-148
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-05211

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05211
Availability details for
Quantity
  • Availability
  • Add to Cart