ENA018108516 Enamine

2-(4-Chloro-benzenesulfonylamino)-3-(1H-indol-3-yl)-propionic acid

  • MDL number MFCD00709349

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05215

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05215
Availability details for
Quantity
  • Availability
  • Add to Cart