ENA018108517 Enamine

2-(2-chloroacetyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

  • MDL number MFCD04606696
  • Cas Number : 1009185-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-05216

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05216
Availability details for
Quantity
  • Availability
  • Add to Cart