ENA018108525 Enamine

11-methyl-13-sulfanyl-1,8-diazatricyclo[7.4.0.0,2,7]trideca-2(7),3,5,8,10,12-hexaene-10-carbonitrile

  • MDL number MFCD02955767
  • Cas Number : 496970-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200-201
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05224

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05224
Availability details for
Quantity
  • Availability
  • Add to Cart