ENA018108534 Enamine

2-Phenyl-pentanoic acid

  • MDL number MFCD03970712
  • Cas Number : 5558-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64...66
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05233

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05233
Availability details for
Quantity
  • Availability
  • Add to Cart