ENA018108550 Enamine

2-(2-Cyano-acetylamino)-benzoic acid

  • MDL number MFCD00088504
  • Cas Number : 87613-40-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05252

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05252
Availability details for
Quantity
  • Availability
  • Add to Cart