ENA018108558 Enamine

2-fluoro-N-(4-hydroxyphenyl)-N-methylbenzene-1-sulfonamide

  • MDL number MFCD03984469
  • Cas Number : 565172-42-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05260

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05260
Availability details for
Quantity
  • Availability
  • Add to Cart