ENA018108571 Enamine

2-Chloro-1-(1,2,5-trimethyl-1H-pyrrol-3-yl)-ethanone

  • MDL number MFCD03970823
  • Cas Number : 565195-14-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81...83
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05274

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05274
Availability details for
Quantity
  • Availability
  • Add to Cart