ENA018108578 Enamine

4-Ureido-benzoic acid

  • MDL number MFCD00025432
  • Cas Number : 6306-25-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 296...298
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05282

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05282
Availability details for
Quantity
  • Availability
  • Add to Cart