ENA018108597 Enamine

4-methoxy-2-[1-(4-methylbenzenesulfonyl)-1H-pyrazole-4-carbonyl]phenol

  • MDL number MFCD03986436
  • Cas Number : 879351-76-5
  • Molecular Weight : 372.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-05303

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05303
Availability details for
Quantity
  • Availability
  • Add to Cart