ENA018108600 Enamine

1-Benzyl-5-(5-fluoro-2-hydroxy-benzoyl)-2-oxo-1,2-dihydro-pyridine-3-carbonitrile

  • MDL number MFCD03973162
  • Cas Number : 730981-96-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05306

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05306
Availability details for
Quantity
  • Availability
  • Add to Cart