ENA018108603 Enamine

ethyl 2-amino-4-(2-methylpropyl)thiophene-3-carboxylate

  • MDL number MFCD03984483
  • Cas Number : 72965-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 31-32
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05309

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05309
Availability details for
Quantity
  • Availability
  • Add to Cart