ENA018108608 Enamine

propan-2-yl 2-(2-chloroacetamido)-4,5,6,7-tetrahydro-1-benzothiophene-3-carboxylate

  • MDL number MFCD03385896
  • Cas Number : 560080-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05314

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05314
Availability details for
Quantity
  • Availability
  • Add to Cart