ENA018108624 Enamine

2-(2,4,6-trimethylphenoxy)acetic acid

  • MDL number MFCD00224887
  • Cas Number : 13333-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05332

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05332
Availability details for
Quantity
  • Availability
  • Add to Cart