ENA018108630 Enamine

4-Isopropoxy-3-methoxy-benzaldehyde

  • MDL number MFCD00837588
  • Cas Number : 2538-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05338

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05338
Availability details for
Quantity
  • Availability
  • Add to Cart