ENA018108633 Enamine

4-(benzyloxy)-3-bromo-5-methoxybenzoic acid

  • MDL number MFCD03973123
  • Cas Number : 568553-88-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05341

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05341
Availability details for
Quantity
  • Availability
  • Add to Cart