ENA018108637 Enamine

ethyl 2-(2-chloroacetamido)-4-(2-methylpropyl)thiophene-3-carboxylate

  • MDL number MFCD03982637
  • Cas Number : 554423-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05345

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05345
Availability details for
Quantity
  • Availability
  • Add to Cart