ENA018108644 Enamine

1,3,4-thiadiazole-2,5-diamine

  • MDL number MFCD00043511
  • Cas Number : 2937-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 197-200
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05352
Availability details for
Quantity
  • Availability
  • Add to Cart