ENA018108664 Enamine

1-(3,4-dichlorophenyl)-3-methyl-1H-pyrazol-5-amine

  • MDL number MFCD03970014
  • Cas Number : 76606-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05372

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05372
Availability details for
Quantity
  • Availability
  • Add to Cart