ENA018108672 Enamine

1-benzoyl-4-(phenylmethylidene)-2-sulfanyl-4,5-dihydro-1H-imidazol-5-one

  • MDL number MFCD03984482
  • Cas Number : 7245-67-2
  • Molecular Weight : 308.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-05380

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05380
Availability details for
Quantity
  • Availability
  • Add to Cart