ENA018108673 Enamine

(1-Phenyl-ethylamino)-acetic acid hydrazide

  • MDL number MFCD03973219
  • Cas Number : 56720-93-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05381

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05381
Availability details for
Quantity
  • Availability
  • Add to Cart