ENA018108678 Enamine

1-(2-chloroacetyl)pyrrolidin-2-one

  • MDL number MFCD03973204
  • Cas Number : 43170-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50-51
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05387

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05387
Availability details for
Quantity
  • Availability
  • Add to Cart