ENA018108708 Enamine

4-(6-Ethoxy-benzothiazol-2-yl)-phenylamine

  • MDL number MFCD02704199
  • Cas Number : 328109-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 203...205
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05419

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05419
Availability details for
Quantity
  • Availability
  • Add to Cart