ENA018108723 Enamine

2-chloro-1-(7-ethyl-1H-indol-3-yl)ethan-1-one

  • MDL number MFCD03984487
  • Cas Number : 571153-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 120-121
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-05437

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05437
Availability details for
Quantity
  • Availability
  • Add to Cart