ENA018108729 Enamine

(3-Methyl-butyl)-urea

  • MDL number MFCD01675989
  • Cas Number : 628-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94...96
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05443

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05443
Availability details for
Quantity
  • Availability
  • Add to Cart