ENA018108736 Enamine

1-[3-(chloromethyl)-4-ethoxyphenyl]ethan-1-one

  • MDL number MFCD01924447
  • Cas Number : 78927-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151-152
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-05452

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05452
Availability details for
Quantity
  • Availability
  • Add to Cart