ENA018108740 Enamine

4-formyl-2-methoxyphenyl 4-methylbenzene-1-sulfonate

  • MDL number MFCD01935657
  • Cas Number : 246224-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05457

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05457
Availability details for
Quantity
  • Availability
  • Add to Cart