ENA018108744 Enamine

7-methyl-2-(trifluoromethyl)-[1,2,4]triazolo[1,5-a]pyrimidin-5-amine

  • MDL number MFCD03976130
  • Cas Number : 554408-52-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-05461

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05461
Availability details for
Quantity
  • Availability
  • Add to Cart