ENA018108746 Enamine

5-(1,3-dioxaindan-5-yl)-4-phenyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD03976138
  • Cas Number : 61066-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05463

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05463
Availability details for
Quantity
  • Availability
  • Add to Cart