ENA018108749 Enamine

6-chloro-4-(chloromethyl)-7-methyl-2H-chromen-2-one

  • MDL number MFCD03976182
  • Cas Number : 41295-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05466

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05466
Availability details for
Quantity
  • Availability
  • Add to Cart