ENA018108760 Enamine

1-(3-methylphenyl)-1H-1,2,3,4-tetrazole-5-thiol

  • MDL number MFCD02073811
  • Cas Number : 41401-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132-133
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05477

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05477
Availability details for
Quantity
  • Availability
  • Add to Cart