ENA018108776 Enamine

1-(2,3-dihydro-1,4-benzodioxine-6-sulfonyl)-4-hydroxypyrrolidine-2-carboxylic acid

  • MDL number MFCD03976220
  • Cas Number : 1008664-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05493

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05493
Availability details for
Quantity
  • Availability
  • Add to Cart