ENA018108788 Enamine

(5-Methyl-4,5-dihydro-thiazol-2-yl)-m-tolyl-amine

  • MDL number MFCD00275292
  • Cas Number : 254732-40-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05507

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05507
Availability details for
Quantity
  • Availability
  • Add to Cart