ENA018108793 Enamine

2-chloro-N-(1,1-dioxo-1lambda6-thiolan-3-yl)acetamide

  • MDL number MFCD03965857
  • Cas Number : 5612-41-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89-98
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-05512

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05512
Availability details for
Quantity
  • Availability
  • Add to Cart