ENA018108799 Enamine

2-(5,6-dichloro-1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid

  • MDL number MFCD03162725
  • Cas Number : 111187-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05518

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05518
Availability details for
Quantity
  • Availability
  • Add to Cart