ENA018108800 Enamine

5-nitro-2-[(1-phenyl-1H-1,2,3,4-tetrazol-5-yl)sulfanyl]benzoic acid

  • MDL number MFCD00514454
  • Cas Number : 309726-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200-201
Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05519

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05519
Availability details for
Quantity
  • Availability
  • Add to Cart