ENA018108803 Enamine

2-Amino-4,5,6,7-tetrahydro-benzo[b]thiophene-3-carboxylic acid methyl ester

  • MDL number MFCD00130099
  • Cas Number : 108354-78-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127...129
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05522

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-05522
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart