ENA018108807 Enamine

N-(6-amino-1-benzyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-5-yl)-N-butyl-2-chloroacetamide

  • MDL number MFCD03984453
  • Cas Number : 568557-43-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05526

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05526
Availability details for
Quantity
  • Availability
  • Add to Cart