ENA018108808 Enamine

6-amino-5-[(2-methoxyethyl)amino]-1-(2-methylpropyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD03986434
  • Cas Number : 554427-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05527

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05527
Availability details for
Quantity
  • Availability
  • Add to Cart