ENA018108810 Enamine

2-chloro-N-[4-(pyrrolidine-1-sulfonyl)phenyl]propanamide

  • MDL number MFCD03984455
  • Cas Number : 554424-71-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05533

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05533
Availability details for
Quantity
  • Availability
  • Add to Cart