ENA018108823 Enamine

3-phenyl-2-sulfanyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD03978460
  • Cas Number : 315707-93-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05549
Availability details for
Quantity
  • Availability
  • Add to Cart