ENA018108827 Enamine

3-ethyl-5-phenyl-2-sulfanyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD03978456
  • Cas Number : 132605-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-05553

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05553
Availability details for
Quantity
  • Availability
  • Add to Cart