ENA018108832 Enamine

6-ethyl-3-methyl-2-sulfanyl-3H,4H-thieno[2,3-d]pyrimidin-4-one

  • MDL number MFCD03978454
  • Cas Number : 733015-18-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05558

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05558
Availability details for
Quantity
  • Availability
  • Add to Cart