ENA018108834 Enamine

N-[(2-chloro-5-nitrophenyl)methylidene]hydroxylamine

  • Cas Number : 89692-57-9
  • Molecular Weight : 200.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 176-177
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05560

Safety Information

signalword Warning
pcodes P264,P270,P273,P301+P312,P330,P391,P501
MSDS https://www.enaminestore.com/catalog/EN300-05560
pictograms GHS07,GHS09
hstatements H302,H411
Availability details for
Quantity
  • Availability
  • Add to Cart