ENA018108835 Enamine

3-nitro-4-(propan-2-yl)benzaldehyde

  • MDL number MFCD03820045
  • Cas Number : 130766-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-05562

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05562
Availability details for
Quantity
  • Availability
  • Add to Cart