ENA018108837 Enamine

1-[(2,6-dichlorophenyl)methyl]-3-methyl-1H-thieno[2,3-c]pyrazole-5-carboxylic acid

  • MDL number MFCD03988518
  • Cas Number : 554439-41-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05564

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05564
Availability details for
Quantity
  • Availability
  • Add to Cart