ENA018108851 Enamine

2-chloro-1-[1-(2-fluorophenyl)-2,5-dimethyl-1H-pyrrol-3-yl]ethan-1-one

  • MDL number MFCD03978337
  • Cas Number : 565170-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80-81
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05581

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05581
Availability details for
Quantity
  • Availability
  • Add to Cart