ENA018108869 Enamine

2-chloro-N-(2-{5-[(2-fluorophenyl)methylidene]-2,4-dioxo-1,3-thiazolidin-3-yl}ethyl)acetamide

  • Cas Number : 568554-60-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05610

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05610
Availability details for
Quantity
  • Availability
  • Add to Cart