ENA018108878 Enamine

3-(2-chloroacetyl)-1-(3-methylbutyl)urea

  • MDL number MFCD03983040
  • Cas Number : 737812-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95-96
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-05619

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05619
Availability details for
Quantity
  • Availability
  • Add to Cart